Assay Details
| Gene Symbol: Pparg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244281.1 | NP_001231210.1 | 5 - 6 | 773 | 74 | |
| XM_007647864.1 | XP_007646054.1 | 5 - 6 | 806 | 74 | ||
| XM_007647865.1 | XP_007646055.1 | 5 - 6 | 787 | 74 | ||
| XM_007647866.1 | XP_007646056.1 | 5 - 6 | 878 | 74 | ||
| XM_007647867.1 | XP_007646057.1 | 5 - 6 | 755 | 74 | ||
| XM_007647868.1 | XP_007646058.1 | 5 - 6 | 812 | 74 | ||
| GenBank mRNA | JI891557.1 | - | 1 - 2 | 194 | 74 | |
| JP055908.1 | - | 1 - 2 | 193 | 74 | ||
| Z30972.1 | - | 5 - 6 | 773 | 74 | ||
More Information
Set Membership: |
|

Pparg
Genomic Map