Assay Details
| Gene Symbol: Fam21c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244293.1 | NP_001231222.1 | 5 - 6 | 545 | 57 | |
| GenBank mRNA | AF371373.1 | - | 5 - 6 | 545 | 57 | |
| JI885163.1 | - | 4 - 5 | 523 | 57 | ||
| JP053502.1 | - | 4 - 5 | 675 | 57 | ||
More Information
Set Membership: |
|

Fam21c
Genomic Map