Assay Details
| Gene Symbol: Dpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244535.1 | NP_001231464.1 | 1 - 2 | 102 | 87 | |
| XM_007647721.1 | XP_007645911.1 | 2 - 3 | 270 | 87 | ||
| GenBank mRNA | AF115410.1 | - | 1 - 2 | 102 | 87 | |
| JI889142.1 | - | 1 - 2 | 393 | 87 | ||
| JP038366.1 | - | 1 - 2 | 393 | 87 | ||
More Information
Set Membership: |
|

Dpm2
Genomic Map