Assay Details
| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246692.1 | NP_001233621.1 | 1 - 2 | 104 | 67 | |
| GenBank mRNA | AF351131.1 | - | 1 - 2 | 104 | 67 | |
More Information
Set Membership: |
|

Ugcg
Genomic Map