Assay Details
| Gene Symbol: B4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246698.1 | NP_001233627.1 | 8 - 9 | 1000 | 99 | |
| GenBank mRNA | AF506839.1 | - | 8 - 9 | 1000 | 99 | |
More Information
Set Membership: |
|

B4galt6
Genomic Map