Assay Details
| Gene Symbol: Htr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246754.1 | NP_001233683.1 | 1 | 1114 | 69 | |
| XM_007648028.1 | XP_007646218.1 | 2 | 1716 | 69 | ||
| XM_007648029.1 | XP_007646219.1 | 2 | 1741 | 69 | ||
| GenBank mRNA | X86458.1 | - | 1 | 1114 | 69 | |
More Information
Set Membership: |
|

Htr1b
Genomic Map