Assay Details
| Gene Symbol: Bcl2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246798.1 | NP_001233727.1 | 1 - 2 | 601 | 64 | |
| XM_007642906.1 | XP_007641096.1 | 2 - 3 | 1040 | 64 | ||
| XM_007642907.1 | XP_007641097.1 | 2 - 3 | 834 | 64 | ||
| XM_007642908.1 | XP_007641098.1 | 2 - 3 | 791 | 64 | ||
| XM_007642909.1 | XP_007641099.1 | 2 - 3 | 905 | 64 | ||
| XM_007642910.1 | XP_007641100.1 | 2 - 3 | 833 | 64 | ||
| GenBank mRNA | EU596590.1 | - | 1 - 2 | 601 | 64 | |
| JP056986.1 | - | 2 - 3 | 756 | 64 | ||
More Information
Set Membership: |
|

Bcl2l1
Genomic Map