Assay Details
| Gene Symbol: Hsp90aa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246821.1 | NP_001233750.1 | 7 - 8 | 1519 | 81 | |
| GenBank mRNA | JI879572.1 | - | 4 - 5 | 829 | 81 | |
| JP066067.1 | - | 3 - 4 | 508 | 81 | ||
| L33676.1 | - | 7 - 8 | 1519 | 81 | ||
More Information
Set Membership: |
|

Hsp90aa1
Genomic Map