Assay Details
| Gene Symbol: Hsp90aa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246821.1 | NP_001233750.1 | 8 - 9 | 1783 | 90 | |
| GenBank mRNA | JI879572.1 | - | 5 - 6 | 1093 | 90 | |
| JP066067.1 | - | 4 - 5 | 772 | 90 | ||
| L33676.1 | - | 8 - 9 | 1783 | 90 | ||
More Information
Set Membership: |
|

Hsp90aa1
Genomic Map