Assay Details
| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246822.1 | NP_001233751.1 | 11 - 12 | 1320 | 61 | |
| XM_007652642.1 | XP_007650832.1 | 12 - 13 | 1434 | 61 | ||
| GenBank mRNA | J04209.1 | - | 11 - 12 | 1320 | 61 | |
| JI878814.1 | - | 2 - 3 | 289 | 61 | ||
| JP055329.1 | - | 2 - 3 | 289 | 61 | ||
More Information
Set Membership: |
|

Impdh2
Genomic Map