Assay Details
| Gene Symbol: Sephs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256868.1 | NP_001243797.1 | 1 | 1317 | 76 | |
| GenBank mRNA | JI889277.1 | - | 1 | 906 | 76 | |
| JP058470.1 | - | 1 | 906 | 76 | ||
More Information
Set Membership: |
|

Sephs2
Genomic Map