Assay Details
| Gene Symbol: Vkorc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265617.2 | NP_001252546.1 | 1 - 2 | 175 | 57 | |
| GenBank mRNA | JI885910.1 | - | 1 - 2 | 204 | 57 | |
| JP054855.1 | - | 1 - 2 | 204 | 57 | ||
| JQ400047.1 | - | 1 - 2 | 180 | 57 | ||
More Information
Set Membership: |
|

Vkorc1
Genomic Map