Assay Details
| Gene Symbol: Oaz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291938.1 | NP_001278867.1 | 4 - 5 | 638 | 75 | |
| XM_007653334.1 | XP_007651524.1 | 5 - 6 | 647 | 75 | ||
More Information
Set Membership: |
|

Oaz3
Genomic Map