Assay Details
| Gene Symbol: Rps14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244519.1 | NP_001231448.1 | 2 - 3 | 183 | 117 | |
| XM_007646826.1 | XP_007645016.1 | 1 - 2 | 258 | 117 | ||
| GenBank mRNA | JI869879.1 | - | 2 - 3 | 232 | 117 | |
| JP040329.1 | - | 2 - 3 | 232 | 117 | ||
| M11241.1 | - | 2 - 3 | 183 | 117 | ||
More Information
Set Membership: |
|

Rps14
Genomic Map