Assay Details
| Gene Symbol: Actb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244575.1 | NP_001231504.1 | 1 - 2 | 119 | 67 | |
| XM_007648665.1 | XP_007646855.1 | 2 - 3 | 218 | 67 | ||
| GenBank mRNA | JI878458.1 | - | 2 - 3 | 213 | 67 | |
| JP045094.1 | - | 2 - 3 | 227 | 67 | ||
More Information
Set Membership: |
|

Actb
Genomic Map