Assay Details
| Gene Symbol: Actb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244575.1 | NP_001231504.1 | 2 - 3 | 357 | 113 | |
| XM_007648665.1 | XP_007646855.1 | 3 - 4 | 456 | 113 | ||
| GenBank mRNA | JI878458.1 | - | 3 - 4 | 451 | 113 | |
| JP040858.1 | - | 1 - 2 | 47 | 113 | ||
| JP045094.1 | - | 3 - 4 | 465 | 113 | ||
More Information
Set Membership: |
|

Actb
Genomic Map