Assay Details
| Gene Symbol: Fut6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244087.1 | NP_001231016.1 | 2 | 1621 | 97 | |
| XM_007642396.1 | XP_007640586.1 | 2 | 1621 | 97 | ||
| GenBank mRNA | AF090449.1 | - | 2 | 1621 | 97 | |
More Information
Set Membership: |
|

Fut6b
Genomic Map