Assay Details
| Gene Symbol: Sumo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244144.1 | NP_001231073.1 | 1 | 414 | 97 | |
| GenBank mRNA | JI862782.1 | - | 1 | 505 | 97 | |
| JP054697.1 | - | 1 | 505 | 97 | ||
| L79950.1 | - | 1 | 414 | 97 | ||
More Information
Set Membership: |
|

Sumo2
Genomic Map