Assay Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501892.2 | XP_003501940.1 | 3 - 4 | 581 | 58 | |
| GenBank mRNA | JI863728.1 | - | 3 - 4 | 368 | 58 | |
More Information
Set Membership: |
|

Dhps
Genomic Map