Assay Details
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506207.2 | XP_003506255.1 | 6 - 7 | 716 | 66 | |
GenBank mRNA | JI878540.1 | - | 6 - 7 | 716 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
