Assay Details
| Gene Symbol: Zscan21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510415.2 | XP_003510463.1 | 2 - 3 | 619 | 67 | |
| XM_007651030.1 | XP_007649220.1 | 2 - 3 | 557 | 67 | ||
| XM_007651031.1 | XP_007649221.1 | 2 - 3 | 759 | 67 | ||
| XM_007651032.1 | XP_007649222.1 | 2 - 3 | 600 | 67 | ||
| GenBank mRNA | JI881917.1 | - | 1 - 2 | 66 | 67 | |
| JP047679.1 | - | 1 - 2 | 66 | 67 | ||
More Information
Set Membership: |
|

Zscan21
Genomic Map