Assay Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514743.2 | XP_003514791.1 | 2 - 3 | 341 | 72 | |
| XM_007654792.1 | XP_007652982.1 | 3 - 4 | 391 | 72 | ||
| XM_007654793.1 | XP_007652983.1 | 3 - 4 | 388 | 72 | ||
| XM_007654796.1 | XP_007652986.1 | 2 - 3 | 338 | 72 | ||
More Information
Set Membership: |
|

Szrd1
Genomic Map