Assay Details
| Gene Symbol: LOC100765482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654040.1 | XP_007652230.1 | 2 - 3 | 224 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100765482| Gene Symbol: LOC100765482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654040.1 | XP_007652230.1 | 2 - 3 | 224 | 100 | |