Assay Details
| Gene Symbol: Sh2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502898.1 | XP_003502946.1 | 4 - 5 | 464 | 64 | |
| XM_003502899.1 | XP_003502947.1 | 4 - 5 | 464 | 64 | ||
More Information
Set Membership: |
|

Sh2d2a
Genomic Map