Assay Details
Gene Symbol: LOC100774966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503515.2 | XP_003503563.1 | 1 - 2 | 476 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
