Assay Details
Gene Symbol: LOC100774966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503515.2 | XP_003503563.1 | 3 - 4 | 952 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
