Assay Details
| Gene Symbol: Alkbh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503527.2 | XP_003503575.2 | 2 - 3 | 608 | 70 | |
| GenBank mRNA | JI892995.1 | - | 1 - 2 | 152 | 70 | |
More Information
Set Membership: |
|

Alkbh2
Genomic Map