Assay Details
| Gene Symbol: C4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508495.2 | XP_003508543.1 | 9 - 10 | 1043 | 50 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4a| Gene Symbol: C4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508495.2 | XP_003508543.1 | 9 - 10 | 1043 | 50 | |
Set Membership: |
|