Assay Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510661.2 | XP_003510709.2 | 9 - 10 | 1541 | 59 | |
| GenBank mRNA | JP063482.1 | - | 3 - 4 | 707 | 59 | |
More Information
Set Membership: |
|

Tmem57
Genomic Map