Assay Details
| Gene Symbol: Myoz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511479.2 | XP_003511527.1 | 5 - 6 | 612 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myoz2| Gene Symbol: Myoz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511479.2 | XP_003511527.1 | 5 - 6 | 612 | 79 | |
Set Membership: |
|