Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653343.1 | XP_007651533.1 | 3 - 4 | 600 | 69 | |
| GenBank mRNA | JI890111.1 | - | 3 - 4 | 600 | 69 | |
More Information
Set Membership: |
|

Capza1
Genomic Map