Assay Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646499.1 | XP_007644689.1 | 1 - 2 | 163 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab27a| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646499.1 | XP_007644689.1 | 1 - 2 | 163 | 65 | |
Set Membership: |
|