Assay Details
| Gene Symbol: Rfx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495029.2 | XP_003495077.1 | 6 - 7 | 625 | 65 | |
| XM_007647342.1 | XP_007645532.1 | 5 - 6 | 614 | 65 | ||
More Information
Set Membership: |
|

Rfx7
Genomic Map