Assay Details
| Gene Symbol: Psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504658.2 | XP_003504706.1 | 4 - 5 | 857 | 56 | |
| GenBank mRNA | JI872433.1 | - | 10 - 11 | 56 | ||
More Information
Set Membership: |
|

Psmc3ip
Genomic Map