Assay Details
| Gene Symbol: Nt5c3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504902.2 | XP_003504950.1 | 5 - 6 | 352 | 82 | |
| XM_007645384.1 | XP_007643574.1 | 5 - 6 | 462 | 82 | ||
| XM_007645386.1 | XP_007643576.1 | 6 - 7 | 394 | 82 | ||
| GenBank mRNA | JI883733.1 | - | 4 - 5 | 399 | 82 | |
More Information
Set Membership: |
|

Nt5c3a
Genomic Map