Assay Details
| Gene Symbol: Pold3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511690.2 | XP_003511738.1 | 9 - 10 | 1085 | 66 | |
| XM_007652227.1 | XP_007650417.1 | 9 - 10 | 1016 | 66 | ||
| XM_007652228.1 | XP_007650418.1 | 9 - 10 | 997 | 66 | ||
| XM_007652229.1 | XP_007650419.1 | 9 - 10 | 979 | 66 | ||
| GenBank mRNA | JI876334.1 | - | 9 - 10 | 1061 | 66 | |
| JP047835.1 | - | 9 - 10 | 1060 | 66 | ||
More Information
Set Membership: |
|

Pold3
Genomic Map