Assay Details
| Gene Symbol: Kansl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515211.2 | XP_003515259.1 | 2 - 3 | 327 | 62 | |
| XM_007655203.1 | XP_007653393.1 | 2 - 3 | 337 | 62 | ||
| XM_007655204.1 | XP_007653394.1 | 1 - 2 | 351 | 62 | ||
| XM_007655207.1 | XP_007653397.1 | 1 - 2 | 203 | 62 | ||
| GenBank mRNA | JI879095.1 | - | 1 - 2 | 262 | 62 | |
| JP051263.1 | - | 1 - 2 | 262 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007655207.1 transcripts.
Set Membership: |
|

Kansl2
Genomic Map