Assay Details
| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650149.1 | XP_007648339.1 | 5 - 6 | 855 | 80 | |
| XM_007650150.1 | XP_007648340.1 | 5 - 6 | 1713 | 80 | ||
More Information
Set Membership: |
|

Prss22
Genomic Map