Assay Details
| Gene Symbol: LOC100756860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649857.1 | XP_007648047.1 | 8 - 9 | 889 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100756860| Gene Symbol: LOC100756860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649857.1 | XP_007648047.1 | 8 - 9 | 889 | 75 | |
Set Membership: |
|