Assay Details
| Gene Symbol: Cpne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641844.1 | XP_007640034.1 | 11 - 12 | 1097 | 58 | |
| XM_007641845.1 | XP_007640035.1 | 10 - 11 | 1015 | 58 | ||
| XM_007641846.1 | XP_007640036.1 | 10 - 11 | 881 | 58 | ||
| XM_007641847.1 | XP_007640037.1 | 10 - 11 | 951 | 58 | ||
| XM_007641848.1 | XP_007640038.1 | 12 - 13 | 4109 | 58 | ||
| GenBank mRNA | JI891658.1 | - | 9 - 10 | 861 | 58 | |
| JP047531.1 | - | 9 - 10 | 861 | 58 | ||
More Information
Set Membership: |
|

Cpne1
Genomic Map