Assay Details
| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655129.1 | XP_007653319.1 | 3 - 4 | 405 | 61 | |
| GenBank mRNA | JI868658.1 | - | 3 - 4 | 369 | 61 | |
More Information
Set Membership: |
|

Serpinf1
Genomic Map