Assay Details
| Gene Symbol: Prickle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649613.1 | XP_007647803.1 | 4 - 5 | 652 | 58 | |
| XM_007649614.1 | XP_007647804.1 | 4 - 5 | 648 | 58 | ||
| XM_007649615.1 | XP_007647805.1 | 4 - 5 | 483 | 58 | ||
| XM_007649616.1 | XP_007647806.1 | 3 - 4 | 443 | 58 | ||
| XM_007649617.1 | XP_007647807.1 | 3 - 4 | 439 | 58 | ||
| XM_007649618.1 | XP_007647808.1 | 4 - 5 | 602 | 58 | ||
| XM_007649619.1 | XP_007647809.1 | 3 - 4 | 393 | 58 | ||
| XM_007649620.1 | XP_007647810.1 | 4 - 5 | 652 | 58 | ||
More Information
Set Membership: |
|

Prickle4
Genomic Map