Assay Details
| Gene Symbol: Spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497207.2 | XP_003497255.1 | 5 - 6 | 516 | 63 | |
| GenBank mRNA | JI866065.1 | - | 5 - 6 | 436 | 63 | |
| JP045132.1 | - | 5 - 6 | 436 | 63 | ||
More Information
Set Membership: |
|

Spag7
Genomic Map