Assay Details
| Gene Symbol: Rab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498103.2 | XP_003498151.1 | 2 - 3 | 314 | 98 | |
| GenBank mRNA | JI866427.1 | - | 2 - 3 | 297 | 98 | |
More Information
Set Membership: |
|

Rab1a
Genomic Map