Assay Details
| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501355.2 | XP_003501403.1 | 2 - 3 | 186 | 68 | |
| GenBank mRNA | JI886442.1 | - | 2 - 3 | 189 | 68 | |
| JP063999.1 | - | 2 - 3 | 189 | 68 | ||
More Information
Set Membership: |
|

Mrpl27
Genomic Map