Assay Details
| Gene Symbol: Tomm40l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506872.2 | XP_003506920.1 | 9 - 10 | 948 | 60 | |
| XM_007647407.1 | XP_007645597.1 | 9 - 10 | 1048 | 60 | ||
| GenBank mRNA | JI878051.1 | - | 6 - 7 | 538 | 60 | |
| JP062578.1 | - | 6 - 7 | 538 | 60 | ||
More Information
Set Membership: |
|

Tomm40l
Genomic Map