Assay Details
| Gene Symbol: Ivd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512596.2 | XP_003512644.1 | 2 - 3 | 268 | 61 | |
| GenBank mRNA | JI872466.1 | - | 2 - 3 | 261 | 61 | |
| JP051863.1 | - | 2 - 3 | 261 | 61 | ||
More Information
Set Membership: |
|

Ivd
Genomic Map