Assay Details
| Gene Symbol: Magoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495730.2 | XP_003495778.1 | 1 - 2 | 209 | 71 | |
| GenBank mRNA | JI873895.1 | - | 1 - 2 | 183 | 71 | |
| JP040696.1 | - | 1 - 2 | 183 | 71 | ||
More Information
Set Membership: |
|

Magoh
Genomic Map