Assay Details
| Gene Symbol: Dnajb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496433.2 | XP_003496481.1 | 2 - 3 | 1263 | 71 | |
| XM_007650117.1 | XP_007648307.1 | 3 - 4 | 1628 | 71 | ||
| GenBank mRNA | JI874889.1 | - | 2 - 3 | 814 | 71 | |
| JP058887.1 | - | 2 - 3 | 814 | 71 | ||
More Information
Set Membership: |
|

Dnajb4
Genomic Map