Assay Details
| Gene Symbol: Nup35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500415.2 | XP_003500463.1 | 3 - 4 | 409 | 68 | |
| GenBank mRNA | JI865990.1 | - | 3 - 4 | 402 | 68 | |
| JP063276.1 | - | 3 - 4 | 402 | 68 | ||
More Information
Set Membership: |
|

Nup35
Genomic Map